Learn More
Perkin Elmer US LLC Yellow PVDF Syringe Filters, 0.45 µm, 13 mm (100/pack)
Supplier:  Perkin Elmer US LLC 02542783
Encompass_Preferred
PVDF membranes are not recommended for use with acetone, DMF, DMSO, or bases >6N. All our syringe filters are HPLC certified. Housing Material: Polypropylene Effective Filtration Area (cm2): 1.09 Pore Size (µm): 0.45 µm Holdup Volume (µl): <25 Sample Volume (mL): <10 Inlet Connection: Female luer lock Outlet Connection: Male luer slip Maximum Operating Temperature: 50°C Maximum Operating Pressure: 87 psi (6.0 bar) at 20 °C Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis Purified sampleproper column operation
Catalog No. 50-211-3347