Learn More
Bioworld YELLOW SEPAROPOREAGAROSE)
Supplier:  Bioworld 201810981
Encompass
This item has a minimum order quantity of 2 per supplier requirements. YELLOW SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consists of Reactive yellow-3 bound to cross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked agarose beads, 4% Dye:Reactive Blue 3 Ex...
Catalog No. 50-197-913