Learn More
Bioworld YELLOW SEPAROPOREAGAROSE)
Catalog No. 50197914
Encompass
YELLOW SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consists of Reactive yellow-3 bound to cross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked agarose beads, 4% Dye:Reactive Blue 3 Ex...
Catalog No. 50-197-914 Supplier Bioworld Supplier No. 201810982
YELLOW SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consists of Reactive yellow-3 bound to cross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked agarose beads, 4% Dye:Reactive Blue 3 Ex...