Learn More
Cole-Parmer Cole-Parmer Solid Color-Coded Silicone Stoppers, Standard Size 12, Yellow; 1/Bag
Catalog No. NC0300548 Shop All Cole-Parmer Products
Encompass_Preferred
Silicone bubbles make these stoppers lightweightthey float on water. Withstand temperatures from 76 to 392>F (60 to 200>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers. Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bags/case.
Catalog No. NC0300548 Supplier Cole-Parmer Supplier No. 0629822
Silicone bubbles make these stoppers lightweightthey float on water. Withstand temperatures from 76 to 392>F (60 to 200>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers. Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bags/case.