Learn More
YSI™ Probe Reconditioning Kit for Model 95 Handheld Dissolved Oxygen Meter
Catalog No. 0932475 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
For Use With (Equipment):
Model 95 Handheld DO Meter
Catalog No. For Use With (Equipment)
09-324-75 Model 95 Handheld DO Meter
1 options
Catalog No. 09-324-75 Supplier YSI™ Supplier No. 9503
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
  • Used to clean sensor surface of YSI Model 95 (13-298-30, 31, 32, 33) MEA probes
  • Includes sanding tool, micro buffing cloth, sanding disc and instructions
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Model 95 Handheld DO Meter