Learn More
Andwin Scientific VWR Tips, 1-200 uL, Yellow, Nonsterile, 96/Hinged Rack, 960/pk
SDP
Catalog No. NC0955512
Encompass_Preferred
Bevel Point Pipet Tips provide maximum sample recovery and minimal fluid retention. They have a beveled end for superior accuracy and reproducibility in microliter volume ranges.
Catalog No. NC0955512 Supplier Andwin Scientific Supplier No. 53508783
Bevel Point Pipet Tips provide maximum sample recovery and minimal fluid retention. They have a beveled end for superior accuracy and reproducibility in microliter volume ranges.