Learn More
Tewes Corporation 4 x 6 4 Mil Vci Flat Bag 1000/Cs
SDP
Catalog No. NC1841701
Encompass_Preferred
VCI Bags (Vapor Corrosion Inhibitors) are easy to use and provide cost effective proven corrosion protection. Ferrous Yellow bags are a special packaging film containing proprietary Zerust corrosion inhibiting additives developed to prevent the corrosion of ferrous metals, such as iron, steel, and cast irons. Bags will protect metals for up to 5 years if properly packed and stored. Offer a shelf life of 2 years if stored at 85 degrees F (30 degrees C) or less, out of direct sunlight and in original packaging. 1000/CASE.
Catalog No. NC1841701 Supplier Tewes Corporation Supplier No. 025F00002B
VCI Bags (Vapor Corrosion Inhibitors) are easy to use and provide cost effective proven corrosion protection. Ferrous Yellow bags are a special packaging film containing proprietary Zerust corrosion inhibiting additives developed to prevent the corrosion of ferrous metals, such as iron, steel, and cast irons. Bags will protect metals for up to 5 years if properly packed and stored. Offer a shelf life of 2 years if stored at 85 degrees F (30 degrees C) or less, out of direct sunlight and in original packaging. 1000/CASE.