Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yellow Wax, Cake, NF, Spectrum™ Chemical
SDP

Catalog No. 18607799 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 lb.
55 lb.
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-799 1 lb. Poly Bottle
18-607-802 55 lb. Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-799 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W10121LB
Add to Cart
Add to Cart

W1012, 8012-89-3

Spectrum™ Chemical Yellow Wax, Cake, NF - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8012-89-3

Specifications

CAS Max % 1
Melting Range 62-65°C
Packaging Poly Bottle
Quantity 1 lb.
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.