Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yellow Wax, Cake, NF, Spectrum™ Chemical
SDP

Catalog No. 18607801 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 lb.
12 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-801 5 lb. Poly Pail
18-607-800 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-801 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W10125LB
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

W1012, 8012-89-3

Spectrum™ Chemical Yellow Wax, Cake, NF - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8012-89-3

Specifications

CAS Max % 1
Melting Range 62°C to 65°C
Packaging Poly Pail
Quantity 5 lb.
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Yellow Wax, Cake
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.