Learn More
ATCC Enterococcus faecium
Catalog No. 502380950
Encompass
Enterococcus faecium strain Vancomycin-dependent #4 is a drug-resistant bacterium that was isolated in 1992 from human blood. This whole-genome sequenced strain is resistant to teicoplanin and vancomycin.
Catalog No. 50-238-0950 Supplier ATCC Supplier No. 51858
Add to Cart
Add to Cart
Enterococcus faecium strain Vancomycin-dependent #4 is a drug-resistant bacterium that was isolated in 1992 from human blood. This whole-genome sequenced strain is resistant to teicoplanin and vancomycin.