Learn More
ATCC Yersinia; enterocolitica; subsp. enterocolitica
Catalog No. 502383740
Encompass
Yersinia enterocolitica subsp. enterocolitica strain 33114 was isolated in New York from a human tissue sample taken from a glanders-like infection on the face. The whole-genome sequenced bacterial type strain can be used in media testing and is recommended as a test microorganism for EP 2.6.27 Microbiological Control of Cellular Products.
Catalog No. 50-238-3740 Supplier ATCC Supplier No. 9610
Yersinia enterocolitica subsp. enterocolitica strain 33114 was isolated in New York from a human tissue sample taken from a glanders-like infection on the face. The whole-genome sequenced bacterial type strain can be used in media testing and is recommended as a test microorganism for EP 2.6.27 Microbiological Control of Cellular Products.