Learn More
Alkali Scientific Yersinia Selective Agar (CIN) 20mL, 10/Pk
SDP
Catalog No. 5000663233
Encompass_Preferred
Yersinia Selective Agar (CIN) is a selective and differential culture medium used for the isolation of Yersinia enterocolitica and related enteric pathogens from clinical and environmental specimens. The formulation contains selective agents that inhibit background microflora while supporting growth of Yersinia species. Characteristic colony morphology, often described as a “bull’s eye” appearance, assists in the visual identification of target organisms during microbiological examination. Each plate contains approximately 20mL of prepared medium in a standard petri dish format for consistent plating performance. This medium is widely used in clinical diagnostics, food safety testing, and environmental microbiology laboratories.
Catalog No. 50-006-63233 Supplier Alkali Scientific Supplier No. YSAP00
Yersinia Selective Agar (CIN) is a selective and differential culture medium used for the isolation of Yersinia enterocolitica and related enteric pathogens from clinical and environmental specimens. The formulation contains selective agents that inhibit background microflora while supporting growth of Yersinia species. Characteristic colony morphology, often described as a “bull’s eye” appearance, assists in the visual identification of target organisms during microbiological examination. Each plate contains approximately 20mL of prepared medium in a standard petri dish format for consistent plating performance. This medium is widely used in clinical diagnostics, food safety testing, and environmental microbiology laboratories.