Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Yersinia Selective Agar
GSA_VA

Catalog No. R01998 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
20 x 25 mL
Format:
12mm x 85mm Monoplate
20mm x 150mm Pour Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01998 10/Pk. 12mm x 85mm Monoplate
R09999 20 x 25 mL 20mm x 150mm Pour Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01998 Supplier Thermo Scientific™ Supplier No. R01998
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples on this prepared media plate after 18 to 24 hours incubation at 32°C.

Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples after 18 to 24 hours using Thermo Scientific™ Yersinia Selective Agar (CIN Agar).

Specifically developed for the optimum growth and recovery of Yersinia enterocolitica, the formulation includes cefsulodin, Irgasan™ and novobiocin. The typical colonies of Yersinia enterocolitica will develop as a red bullseye surrounded by a transparent border and will vary considerably among serotypes in colony size, smoothness and the ratio of the border to center diameter. Most other organisms that are capable of growing will produce larger colonies (>2mm in diameter) with diffuse pinkish centers and opaque outer zones. Serratia liquefaciens, Citrobacter freundii and Enterobacter agglomerans may give a colonial morphology resembling Yersinia enterocolitica. These organisms can be differentiated from Yersinia enterocolitica by biochemical tests.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF.
Description Yersinia Selective Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.