Learn More
Teknova YES Agar Plates. 100mm Plates, 20 Plates/Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387469
Encompass
0.5% Yeast Extract, 3.0% Glucose, 225g/mL Adenine, 225g/mL Histidine, 225g/mL Leucine, 225g/mL Uracil, 225g/mL Lysine, 2.0% Agar
Catalog No. 50-238-7469 Supplier Teknova Supplier No. Y30054PK
0.5% Yeast Extract, 3.0% Glucose, 225g/mL Adenine, 225g/mL Histidine, 225g/mL Leucine, 225g/mL Uracil, 225g/mL Lysine, 2.0% Agar