Learn More
Sino Biological Yellow fever virus (YFV) (strain 17D vaccine) Envelope Protein (His Tag)
Catalog No. 502992981
Encompass_Preferred
A DNA sequence encoding the Yellow fever virus (YFV) (strain 17D vaccine) Envelope Protein (P03314) (Arg126-Arg244) was fused with the N-terminal polyhistidine tag followed by a GST tag.
Catalog No. 50-299-2981 Supplier Sino Biological Supplier No. 41052V53E100
A DNA sequence encoding the Yellow fever virus (YFV) (strain 17D vaccine) Envelope Protein (P03314) (Arg126-Arg244) was fused with the N-terminal polyhistidine tag followed by a GST tag.