Learn More
ENCAPSULITE INTERNATIONAL INC YG10 Roll
Supplier:  ENCAPSULITE INTERNATIONAL INC WFYG10
Encompass
Safelight Filter: YG10 - Yellow GoldSpectral Limit: 500Dimensions (feet): 4x32
Catalog No. NC2683697