Learn More
Medchemexpress LLC YG1702, 10mg | CAS No. : 724737-08-0 | Purity: 99.21%
SDP
Catalog No. NC3713037
Encompass_Preferred
YG1702 is a potent ALDH18A1-specific inhibitor. YG1702 attenuates the growth of MYCN-amplified NB and down-regulates MYCN. YG1702 physically interacts with ALDH18A1 with a high affinity and might potentially affect its enzymatic activity.
Catalog No. NC3713037 Supplier Medchemexpress LLC Supplier No. HY15644310MG
YG1702 is a potent ALDH18A1-specific inhibitor. YG1702 attenuates the growth of MYCN-amplified NB and down-regulates MYCN. YG1702 physically interacts with ALDH18A1 with a high affinity and might potentially affect its enzymatic activity.