Learn More
Sigma Aldrich Fine Chemicals Biosciences YGC Agar | 500g
Catalog No. NC1366998
Encompass
Selective agar for the enumeration and isolation of yeasts and molds in dairy products.
Catalog No. NC1366998 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 95765500G
Selective agar for the enumeration and isolation of yeasts and molds in dairy products.