Learn More
Teknova YGC Agar Plates with Chloramphenicol-100. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387245
Encompass
0.5% Yeast Extract, 2.0% Glucose, 1.5% Agar, 100g/mL Chloramphenicol
Catalog No. 50-238-7245 Supplier Teknova Supplier No. Y15004PK
0.5% Yeast Extract, 2.0% Glucose, 1.5% Agar, 100g/mL Chloramphenicol