Learn More
Medchemexpress LLC Ygsy2p-IN-1 | 2415003-97-1 | 98.6% | 50 MG
SDP
Catalog No. 5000338537
Encompass_Preferred
yGsy2p-IN-1 is a potent inhibitor for yeast glycogen synthase 2 (yGsy2p) and a competitive human glycogen synthase 1 (hGYS1) inhibitor. It is a pyrazole inhibitor used for glycogen storage diseases (GSDs).
  • Potent inhibitor for yeast glycogen synthase 2 (yGsy2p)
  • Competitive human glycogen synthase 1 (hGYS1) inhibitor with an IC50 of 2.75 μM and a Ki of 1.31 μM for wild-type hGYS1
  • Used for glycogen storage diseases (GSDs)
Catalog No. 50-003-38537 Supplier Medchemexpress LLC Supplier No. HY13106250MG
yGsy2p-IN-1 is a potent inhibitor for yeast glycogen synthase 2 (yGsy2p) and a competitive human glycogen synthase 1 (hGYS1) inhibitor. It is a pyrazole inhibitor used for glycogen storage diseases (GSDs).
  • Potent inhibitor for yeast glycogen synthase 2 (yGsy2p)
  • Competitive human glycogen synthase 1 (hGYS1) inhibitor with an IC50 of 2.75 μM and a Ki of 1.31 μM for wild-type hGYS1
  • Used for glycogen storage diseases (GSDs)