Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YH-306 | 1373764-75-0 | 98.3% | 5 MG
SDP

Catalog No. 5000234415
Encompass_Preferred

YH-306 is a small-molecule antitumor agent that inhibits focal adhesion kinase (FAK) signaling and reduces cancer cell migration, invasion, and metastasis in preclinical studies.

  • Inhibits focal adhesion kinase (FAK) signaling and related pathways.
  • Reduces expression of MMP2 and MMP9.
  • Inhibits Arp2/3-mediated actin polymerization.
  • Molecular formula: C19H18N2O2.
  • Molecular weight: 306.36 g/mol.
  • Purity: 98.3%.
  • Appearance: light yellow to light brown solid.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-34415 Supplier Medchemexpress LLC Supplier No. HY1202135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YH-306 is a small-molecule antitumor agent that inhibits focal adhesion kinase (FAK) signaling and reduces cancer cell migration, invasion, and metastasis in preclinical studies.

  • Inhibits focal adhesion kinase (FAK) signaling and related pathways.
  • Reduces expression of MMP2 and MMP9.
  • Inhibits Arp2/3-mediated actin polymerization.
  • Molecular formula: C19H18N2O2.
  • Molecular weight: 306.36 g/mol.
  • Purity: 98.3%.
  • Appearance: light yellow to light brown solid.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.