Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yhhu-3792 | 2097826-24-7 | 99.3% | 50 MG
SDP

Catalog No. 5000234880
Encompass_Preferred

Yhhu-3792 is a small-molecule Notch activator used in neuroscience research to promote neural stem cell self-renewal, increase Hes3 and Hes5 expression, and improve spatial and episodic memory in animal models. It is supplied as a high-purity research reagent with good solubility in DMSO for in vitro studies.

  • Notch activator with reported neurogenic activity.
  • Supports neural stem cell self-renewal and upregulation of Hes3 and Hes5.
  • High purity: 99.34% (analytical grade).
  • Molecular weight 400.47 g/mol; formula C24H24N4O2.
  • Soluble in DMSO (≈66.67 mg/mL); ultrasonic assistance may be required.
  • Store sealed, away from moisture and light; recommended cold storage for solutions.

Catalog No. 50-002-34880 Supplier Medchemexpress LLC Supplier No. HY12078250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yhhu-3792 is a small-molecule Notch activator used in neuroscience research to promote neural stem cell self-renewal, increase Hes3 and Hes5 expression, and improve spatial and episodic memory in animal models. It is supplied as a high-purity research reagent with good solubility in DMSO for in vitro studies.

  • Notch activator with reported neurogenic activity.
  • Supports neural stem cell self-renewal and upregulation of Hes3 and Hes5.
  • High purity: 99.34% (analytical grade).
  • Molecular weight 400.47 g/mol; formula C24H24N4O2.
  • Soluble in DMSO (≈66.67 mg/mL); ultrasonic assistance may be required.
  • Store sealed, away from moisture and light; recommended cold storage for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.