Learn More
TARGETMOL CHEMICALS INC YIL 781 5MG
SDP
Catalog No. 502580862
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YIL 781 is a selective ghrelin receptor antagonist (GHS-R1a) (Ki = 17 nM) showing a weak affinity for kinesin receptors (K = 6 uM). YIL 781 improves glucose homeostasis in vivo and in vitro by blocking ghrelin secretion. purity: 99%
Catalog No. 50-258-0862 Supplier TARGETMOL CHEMICALS INC Supplier No. T235435MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YIL 781 is a selective ghrelin receptor antagonist (GHS-R1a) (Ki = 17 nM) showing a weak affinity for kinesin receptors (K = 6 uM). YIL 781 improves glucose homeostasis in vivo and in vitro by blocking ghrelin secretion. purity: 99%