Learn More
Medchemexpress LLC YIL781 hydrochloride | 1640226-17-0 | 99.48% | 445.96 | 25 MG
SDP
Catalog No. 5000350129
Encompass_Preferred
YIL781 hydrochloride is a potent, orally active ghrelin receptor (GHSR) antagonist. It has been shown to improve glucose homeostasis in rats and effectively inhibits the calcium response induced by ghrelin.
  • Potent and orally active ghrelin receptor (GHSR) antagonist.
  • Produces a greater improvement in glucose homeostasis in rats.
  • Inhibits the calcium response induced by ghrelin with pIC50 values of 7.90 and 8.27.
  • For research use only.
Catalog No. 50-003-50129 Supplier Medchemexpress LLC Supplier No. 25MGHY13964A
YIL781 hydrochloride is a potent, orally active ghrelin receptor (GHSR) antagonist. It has been shown to improve glucose homeostasis in rats and effectively inhibits the calcium response induced by ghrelin.
  • Potent and orally active ghrelin receptor (GHSR) antagonist.
  • Produces a greater improvement in glucose homeostasis in rats.
  • Inhibits the calcium response induced by ghrelin with pIC50 values of 7.90 and 8.27.
  • For research use only.