Learn More
Medchemexpress LLC YIL781 hydrochloride | 1640226-17-0 | 99.5% | 445.96 g/mol | C24H29ClFN3O2 | 5 MG
SDP
Catalog No. 5000222252
Encompass_Preferred
YIL781 hydrochloride is the hydrochloride salt of a potent, orally active ghrelin receptor (GHSR) antagonist used in preclinical research to study ghrelin signaling, metabolic regulation, and insulin secretion. The compound is provided with solubility guidance and storage recommendations for reliable use in experimental workflows.
  • Potent ghrelin receptor (GHSR) antagonist suitable for pharmacological studies.
  • Usable in vitro and in vivo applications.
  • High purity (~99.5%) for reliable experimental results.
  • Soluble in DMSO; formulation guidance provided for aqueous preparations.
  • Stable when stored sealed at -20°C; solvent stability recommendations available.
  • Available in small research pack sizes for early-stage studies.
Catalog No. 50-002-22252 Supplier Medchemexpress LLC Supplier No. HY13964A5MG
YIL781 hydrochloride is the hydrochloride salt of a potent, orally active ghrelin receptor (GHSR) antagonist used in preclinical research to study ghrelin signaling, metabolic regulation, and insulin secretion. The compound is provided with solubility guidance and storage recommendations for reliable use in experimental workflows.
  • Potent ghrelin receptor (GHSR) antagonist suitable for pharmacological studies.
  • Usable in vitro and in vivo applications.
  • High purity (~99.5%) for reliable experimental results.
  • Soluble in DMSO; formulation guidance provided for aqueous preparations.
  • Stable when stored sealed at -20°C; solvent stability recommendations available.
  • Available in small research pack sizes for early-stage studies.