Learn More
Abcam YIL781 hydrochloride, Ghrelin receptor antagonist, 10MG
Catalog No. 502706901
Encompass_Preferred
MW 445.96 g/mol, Purity >99%. Ghrelin receptor (GHS-R1a) antagonist (Ki = 17 nM). Competitive antagonist activity in vitro (pKB = 7.54). Displays no affinity for the motilin receptor (Ki = 6 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6901 Supplier Abcam Supplier No. AB14110610MG
MW 445.96 g/mol, Purity >99%. Ghrelin receptor (GHS-R1a) antagonist (Ki = 17 nM). Competitive antagonist activity in vitro (pKB = 7.54). Displays no affinity for the motilin receptor (Ki = 6 μM).
The product is subject to the following: Abcam Restricted Use Statement