Learn More
Aviva Systems Biology Corp YIPF1 Antibody - C-terminal region (ARP49421_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49421_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YIPF1. It was validated on Western Blot. . Alternate names: DJ167A19.1, FinGER1ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-416