Learn More
Aviva Systems Biology Corp YIPF2 Antibody - middle region (ARP49790_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49790_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against YIPF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human YIPF2. Alternate names: FinGER2, MGC3262ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-450