Learn More
Selleck Chemical LLC YJ1206 E4760-100MG
SDP
Catalog No. 5000039572
Encompass_Preferred
YJ1206 is an orally bioavailable highly specific and potent degrader of CDK12/13 with an IC50 of 12 55 nM in VCaP cells It increases DNA damage induces apoptosis and demonstrates robust anti-tumor activity in vivo
Catalog No. 50-000-39572 Supplier Selleck Chemical LLC Supplier No. E4760100MG
YJ1206 is an orally bioavailable highly specific and potent degrader of CDK12/13 with an IC50 of 12 55 nM in VCaP cells It increases DNA damage induces apoptosis and demonstrates robust anti-tumor activity in vivo