Learn More
Medchemexpress LLC Yk-2168 | 2571068-74-9 | 98.1% | 315.80 | 1 ML
SDP
Catalog No. 5000392088
Encompass_Preferred
YK-2168 is a potent and selective CDK9 inhibitor (IC50 of 5.9 nM) that targets the pS2-RNA Pol II C-terminal domain. It induces apoptosis, suppresses CDK9-regulated genes like MYC and Mcl1, and inhibits tumor growth in CDX mice models. This compound is suitable for cancer research, particularly leukemia.
  • Potent and selective CDK9 inhibitor (IC50 of 5.9 nM).
  • Induces apoptosis and suppresses CDK9-regulated genes.
  • Inhibits tumor growth in CDX mice models (80% inhibition at 10 mg/kg).
  • Highly selective over CDK1 and CDK2, with no hERG channel inhibition up to 30 μM.
  • Reduces pS2-RNA Pol II CTD levels and upregulates cleaved caspase 3.
  • Demonstrates good safety in vivo with no significant body weight changes.
Catalog No. 50-003-92088 Supplier Medchemexpress LLC Supplier No. HY16291910MM/1ML
YK-2168 is a potent and selective CDK9 inhibitor (IC50 of 5.9 nM) that targets the pS2-RNA Pol II C-terminal domain. It induces apoptosis, suppresses CDK9-regulated genes like MYC and Mcl1, and inhibits tumor growth in CDX mice models. This compound is suitable for cancer research, particularly leukemia.
  • Potent and selective CDK9 inhibitor (IC50 of 5.9 nM).
  • Induces apoptosis and suppresses CDK9-regulated genes.
  • Inhibits tumor growth in CDX mice models (80% inhibition at 10 mg/kg).
  • Highly selective over CDK1 and CDK2, with no hERG channel inhibition up to 30 μM.
  • Reduces pS2-RNA Pol II CTD levels and upregulates cleaved caspase 3.
  • Demonstrates good safety in vivo with no significant body weight changes.