Learn More
Medchemexpress LLC YK-2168 | 2571068-74-9 | 98.1% | 315.80 | 25 MG
SDP
Catalog No. 5000395389
Encompass_Preferred
YK-2168 is a potent and selective CDK9 inhibitor with an IC50 of 5.9 nM. It inhibits phosphorylation of the CDK9 substrate pS2-RNA Pol II C-terminal domain. This compound induces apoptosis in tumor cells, suppresses the expression of CDK9-regulated genes, and inhibits tumor growth in CDX mice models. It is suitable for cancer research, including leukemia.
  • Potent and selective CDK9 inhibitor (IC50 = 5.9 nM)
  • Inhibits phosphorylation of the CDK9 substrate pS2-RNA Pol II C-terminal domain
  • Induces apoptosis in tumor cells
  • Suppresses expression of CDK9-regulated genes (e.g., MYC and Mcl1)
  • Inhibits tumor growth in CDX mice models
  • High selectivity over CDK1 and CDK2
  • Does not inhibit hERG channel activity at concentrations up to 30 μM
  • Demonstrates a good safety profile in animal models
Catalog No. 50-003-95389 Supplier Medchemexpress LLC Supplier No. 25MGHY162919
YK-2168 is a potent and selective CDK9 inhibitor with an IC50 of 5.9 nM. It inhibits phosphorylation of the CDK9 substrate pS2-RNA Pol II C-terminal domain. This compound induces apoptosis in tumor cells, suppresses the expression of CDK9-regulated genes, and inhibits tumor growth in CDX mice models. It is suitable for cancer research, including leukemia.
  • Potent and selective CDK9 inhibitor (IC50 = 5.9 nM)
  • Inhibits phosphorylation of the CDK9 substrate pS2-RNA Pol II C-terminal domain
  • Induces apoptosis in tumor cells
  • Suppresses expression of CDK9-regulated genes (e.g., MYC and Mcl1)
  • Inhibits tumor growth in CDX mice models
  • High selectivity over CDK1 and CDK2
  • Does not inhibit hERG channel activity at concentrations up to 30 μM
  • Demonstrates a good safety profile in animal models