Learn More
Medchemexpress LLC YK-3-237 | 1215281-19-8 | 99.5% | 372.18 | 100 MG
SDP
Catalog No. 5000400665
Encompass_Preferred
YK-3-237 is a SIRT1 activator that targets mutant p53. It inhibits the proliferation of triple-negative breast cancer cells.
  • SIRT1 activator
  • Targets mutant p53
  • Inhibits the proliferation of triple-negative breast cancer cells
  • Exhibits anti-proliferative activities towards most breast cancer cell lines at submicromolar concentrations
  • Preferentially inhibits the proliferation of breast cancer cell lines carrying mtp53
  • Deacetylates mtp53 in TNBC cell lines
  • Potent activator of Sirt1, on the activation of renal interstitial fibroblasts using NRK-49F cells
  • Significantly reduces expression of α-SMA and fibronectin in a dose-dependent manner in NRK-49F cells
Catalog No. 50-004-00665 Supplier Medchemexpress LLC Supplier No. HY19634100MG
YK-3-237 is a SIRT1 activator that targets mutant p53. It inhibits the proliferation of triple-negative breast cancer cells.
  • SIRT1 activator
  • Targets mutant p53
  • Inhibits the proliferation of triple-negative breast cancer cells
  • Exhibits anti-proliferative activities towards most breast cancer cell lines at submicromolar concentrations
  • Preferentially inhibits the proliferation of breast cancer cell lines carrying mtp53
  • Deacetylates mtp53 in TNBC cell lines
  • Potent activator of Sirt1, on the activation of renal interstitial fibroblasts using NRK-49F cells
  • Significantly reduces expression of α-SMA and fibronectin in a dose-dependent manner in NRK-49F cells