Learn More
Medchemexpress LLC YK-3-237 | 1215281-19-8 | 99.5% | 372.18 | 5 MG
SDP
Catalog No. 5000401377
Encompass_Preferred
YK-3-237 is a SIRT1 activator that targets mutant p53 and inhibits the proliferation of triple-negative breast cancer cells.
  • SIRT1 activator
  • Targets mutant p53
  • Inhibits the proliferation of triple-negative breast cancer cells
  • Exhibits anti-proliferative activities toward most breast cancer cell lines tested at submicromolar concentrations
  • Preferentially inhibits the proliferation of breast cancer cell lines carrying mtp53
  • Deacetylates mtp53 in TNBC cell lines
  • Potent activator of Sirt1 on the activation of renal interstitial fibroblasts using NRK-49F cells
  • Exposure significantly reduces the expression of α-SMA and fibronectin in a dose-dependent manner
Catalog No. 50-004-01377 Supplier Medchemexpress LLC Supplier No. HY196345MG
YK-3-237 is a SIRT1 activator that targets mutant p53 and inhibits the proliferation of triple-negative breast cancer cells.
  • SIRT1 activator
  • Targets mutant p53
  • Inhibits the proliferation of triple-negative breast cancer cells
  • Exhibits anti-proliferative activities toward most breast cancer cell lines tested at submicromolar concentrations
  • Preferentially inhibits the proliferation of breast cancer cell lines carrying mtp53
  • Deacetylates mtp53 in TNBC cell lines
  • Potent activator of Sirt1 on the activation of renal interstitial fibroblasts using NRK-49F cells
  • Exposure significantly reduces the expression of α-SMA and fibronectin in a dose-dependent manner