Learn More
Medchemexpress LLC YK-3-237 | 1215281-19-8 | 99.5% | 372.18 | 50 MG
SDP
Catalog No. 5000402059
Encompass_Preferred
YK-3-237 is a SIRT1 activator that targets mutant p53 and inhibits the proliferation of triple-negative breast cancer cells. This compound offers a potent and selective approach to cancer research with several key activities:
  • Exhibits anti-proliferative activities toward most breast cancer cell lines at submicromolar concentration.
  • Preferentially inhibits the proliferation of breast cancer cell lines carrying mtp53.
  • Inhibits the proliferation of triple-negative breast cancer (TNBC) cell lines.
  • Deacetylates mtp53 in TNBC cell lines.
  • Potent activator of Sirt1.
Catalog No. 50-004-02059 Supplier Medchemexpress LLC Supplier No. HY1963450MG
YK-3-237 is a SIRT1 activator that targets mutant p53 and inhibits the proliferation of triple-negative breast cancer cells. This compound offers a potent and selective approach to cancer research with several key activities:
  • Exhibits anti-proliferative activities toward most breast cancer cell lines at submicromolar concentration.
  • Preferentially inhibits the proliferation of breast cancer cell lines carrying mtp53.
  • Inhibits the proliferation of triple-negative breast cancer (TNBC) cell lines.
  • Deacetylates mtp53 in TNBC cell lines.
  • Potent activator of Sirt1.