Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC YK-3-237 | 1215281-19-8 | MFCD28167612 | 99.5% | 372.18 | C19H21BO7 | 10 MG
SDP

Catalog No. 5000298363
Encompass_Preferred

YK-3-237 is a research-grade small molecule SIRT1 activator that targets mutant p53 and has demonstrated antiproliferative activity against triple-negative breast cancer cell lines. Supplied in solid form or as a DMSO solution for in vitro research use.

  • Acts as a SIRT1 activator targeting mutant p53.
  • Inhibits proliferation of triple-negative breast cancer cells.
  • Provided as solid and as a DMSO solution for in vitro research.
  • High purity (99.54%) suitable for research applications.
  • Molecular weight 372.18 and chemical formula C19H21BO7 for reference.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C (long-term) or -20°C (short-term).

Catalog No. 50-002-98363 Supplier Medchemexpress LLC Supplier No. HY1963410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YK-3-237 is a research-grade small molecule SIRT1 activator that targets mutant p53 and has demonstrated antiproliferative activity against triple-negative breast cancer cell lines. Supplied in solid form or as a DMSO solution for in vitro research use.

  • Acts as a SIRT1 activator targeting mutant p53.
  • Inhibits proliferation of triple-negative breast cancer cells.
  • Provided as solid and as a DMSO solution for in vitro research.
  • High purity (99.54%) suitable for research applications.
  • Molecular weight 372.18 and chemical formula C19H21BO7 for reference.
  • Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C (long-term) or -20°C (short-term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.