Learn More
Selleck Chemical LLC YK-4-279
SDP
Catalog No. 5000000371
Encompass_Preferred
YK-4-279 is a potent inhibitor of EWS-FLI1 binding to RNA helicase A (RHA)
Catalog No. 50-000-00371 Supplier Selleck Chemical LLC Supplier No. S7679200MG
Add to Cart
Add to Cart
YK-4-279 is a potent inhibitor of EWS-FLI1 binding to RNA helicase A (RHA)