Learn More
Medchemexpress LLC YK-4-279 | 1037184-44-3 | 99.9% | C17H13Cl2NO4 | 1 ML
SDP
Catalog No. 5000356334
Encompass_Preferred
YK-4-279 is an inhibitor that blocks RNA Helicase A (RHA) binding with EWS-FLI1, an oncogenic protein. It induces apoptosis and exhibits anti-proliferation activities against various cancer cells. This compound contains a chiral center and can be separated into two enantiomers, making it suitable for cancer research.
  • Blocks RNA Helicase A (RHA) binding with EWS-FLI1
  • Induces apoptosis
  • Exhibits anti-proliferation activities against various cancer cells
  • Contains a chiral center
  • Can be separated into two enantiomers
  • Used in cancer research
Catalog No. 50-003-56334 Supplier Medchemexpress LLC Supplier No. HY1450710MM/1ML
Add to Cart
Add to Cart
YK-4-279 is an inhibitor that blocks RNA Helicase A (RHA) binding with EWS-FLI1, an oncogenic protein. It induces apoptosis and exhibits anti-proliferation activities against various cancer cells. This compound contains a chiral center and can be separated into two enantiomers, making it suitable for cancer research.
  • Blocks RNA Helicase A (RHA) binding with EWS-FLI1
  • Induces apoptosis
  • Exhibits anti-proliferation activities against various cancer cells
  • Contains a chiral center
  • Can be separated into two enantiomers
  • Used in cancer research