Learn More
Medchemexpress LLC Ykl-05-099 | 1936529-65-5 | 99.8% | 600.11 g/mol | C32H34ClN7O3 | 1 ML
SDP
Catalog No. 5000211441
Encompass_Preferred
YKL-05-099 is a small-molecule inhibitor of salt-inducible kinases (SIK) supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for biochemical and cell-based assays, with reported IC50 values of ≈10 nM (SIK1), ≈40 nM (SIK2), and ≈30 nM (SIK3).
  • Potent inhibition of SIK1-SIK3 in biochemical assays.
  • Ready-to-use 10 mM DMSO stock for convenience.
  • High solubility in DMSO and appreciable PBS solubility for in vitro work.
  • Storage stability data provided for powder and solvent forms.
  • Suitable for kinase profiling, cell-based assays, and immunology studies.
Catalog No. 50-002-11441 Supplier Medchemexpress LLC Supplier No. HY10114710MM 1ML
YKL-05-099 is a small-molecule inhibitor of salt-inducible kinases (SIK) supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is intended for biochemical and cell-based assays, with reported IC50 values of ≈10 nM (SIK1), ≈40 nM (SIK2), and ≈30 nM (SIK3).
  • Potent inhibition of SIK1-SIK3 in biochemical assays.
  • Ready-to-use 10 mM DMSO stock for convenience.
  • High solubility in DMSO and appreciable PBS solubility for in vitro work.
  • Storage stability data provided for powder and solvent forms.
  • Suitable for kinase profiling, cell-based assays, and immunology studies.