Learn More
Medchemexpress LLC YKL-05-099 (5MG)
SDP
Catalog No. NC1626955
Encompass_Preferred
YKL-05-099 is a salt-inducible kinase (SIK) probe; inhibits SIK2 with an IC50 of 40 nM.Purity: 99.76%, CAS 1936529-65-5
Catalog No. NC1626955 Supplier Medchemexpress LLC Supplier No. HY1011475MG
YKL-05-099 is a salt-inducible kinase (SIK) probe; inhibits SIK2 with an IC50 of 40 nM.Purity: 99.76%, CAS 1936529-65-5