Learn More
TARGETMOL CHEMICALS INC YKL-06-061 10MG
SDP
Catalog No. 502576671
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YKL-06-061 is a selective salt-inducible kinase (SIK) inhibitor with IC50 values of 6.56 nM/1.77 nM/20.5 nM for SIK1/2/3 respectively. purity: 99%
Catalog No. 50-257-6671 Supplier TARGETMOL CHEMICALS INC Supplier No. T557610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YKL-06-061 is a selective salt-inducible kinase (SIK) inhibitor with IC50 values of 6.56 nM/1.77 nM/20.5 nM for SIK1/2/3 respectively. purity: 99%