Learn More
Medchemexpress LLC HY-120056 10mg Medchemexpress, YKL-06-061 CAS:2172617-15-9 Purity:>98%
SDP
Catalog No. 501969570
Encompass_Preferred
Medchemexpress, HY-120056 10mg YKL-06-061 CAS:2172617-15-9 YKL-06-061 is a potent, selective, second-generation salt-inducible kinase (SIK) inhibitor with IC50 values of 6.56 nM/1.77 nM/20.5 nM for SIK1/2/3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9570 Supplier Medchemexpress LLC Supplier No. HY12005610MG
Medchemexpress, HY-120056 10mg YKL-06-061 CAS:2172617-15-9 YKL-06-061 is a potent, selective, second-generation salt-inducible kinase (SIK) inhibitor with IC50 values of 6.56 nM/1.77 nM/20.5 nM for SIK1/2/3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.