Learn More
Medchemexpress LLC YKL-1-116 5mg | 1957202-71-9 | 606.72 | C34H38N8O3 | 5 MG
SDP
Catalog No. 5000227235
Encompass_Preferred
YKL-1-116 is a selective, covalent inhibitor of cyclin-dependent kinase 7 (CDK7) used in laboratory research to probe CDK7-dependent cell cycle and transcriptional processes. It is more potent than related inhibitors against CDK7 and shows limited activity against CDK9, CDK12, and CDK13. Supplied as a small, high-purity solid for research use only (not for human consumption).
  • Selective covalent inhibitor of CDK7.
  • Higher potency against CDK7 compared with related compounds.
  • Minimal activity against CDK9, CDK12, and CDK13.
  • High reported purity (~98.7%).
  • Provided as a small solid quantity suitable for in vitro research.
  • Molecular weight 606.72 and formula C34H38N8O3.
Catalog No. 50-002-27235 Supplier Medchemexpress LLC Supplier No. HY1168715MG
YKL-1-116 is a selective, covalent inhibitor of cyclin-dependent kinase 7 (CDK7) used in laboratory research to probe CDK7-dependent cell cycle and transcriptional processes. It is more potent than related inhibitors against CDK7 and shows limited activity against CDK9, CDK12, and CDK13. Supplied as a small, high-purity solid for research use only (not for human consumption).
  • Selective covalent inhibitor of CDK7.
  • Higher potency against CDK7 compared with related compounds.
  • Minimal activity against CDK9, CDK12, and CDK13.
  • High reported purity (~98.7%).
  • Provided as a small solid quantity suitable for in vitro research.
  • Molecular weight 606.72 and formula C34H38N8O3.