Learn More
Medchemexpress LLC HY-101257 10mg , YKL-5-124 CAS:1957203-01-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10125710MG
Encompass_Preferred
HY-101257 10mg YKL-5-124 CAS: 1957203-01-8 YKL-5-124 is a potent, selective, irreversible and covalent CDK7 inhibitor with IC50s of 53.5 nM and 9.7 nM for CDK7 and CDK7/Mat1/CycH, respectively. YKL-5-124 is >100-fold greater selective for CDK7 than CDK9 and CDK2, and inactive against CDK12 and CDK13. YKL-5-124 induces a strong cell-cycle arrest, inhibits E2F-driven gene expression, and exhibits little effect on RNA polymerase II phosphorylation status. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0395