Learn More
Selleck Chemical LLC YKL-5-124 S8863-10MM/1ML
SDP
Catalog No. 5000025389
Encompass_Preferred
YKL-5-124 is a potent selective and covalent CDK7 inhibitor with an IC50 of 9 7 nM 1300 nM and 3020 nM for inhibiting CDK7/Mat1/CycH CDK2 and CDK9 respectively It displays biochemical and cellular selectivity for CDK7 over CDK12/13
Catalog No. 50-000-25389 Supplier Selleck Chemical LLC Supplier No. S886310MM/1ML
YKL-5-124 is a potent selective and covalent CDK7 inhibitor with an IC50 of 9 7 nM 1300 nM and 3020 nM for inhibiting CDK7/Mat1/CycH CDK2 and CDK9 respectively It displays biochemical and cellular selectivity for CDK7 over CDK12/13