Learn More
TARGETMOL CHEMICALS INC YKL-5-124 5MG
SDP
Catalog No. 502580951
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YKL-5-124 a potent selective and covalent CDK7 inhibitor with an IC50 of 9.7 nM 1300 nM and 3020 nM for CDK7/Mat1/CycH CDK2 and CDK9 respectively displays biochemical and cellular selectivity for CDK7 over CDK12/13. purity: 99%
Catalog No. 50-258-0951 Supplier TARGETMOL CHEMICALS INC Supplier No. T224615MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. YKL-5-124 a potent selective and covalent CDK7 inhibitor with an IC50 of 9.7 nM 1300 nM and 3020 nM for CDK7/Mat1/CycH CDK2 and CDK9 respectively displays biochemical and cellular selectivity for CDK7 over CDK12/13. purity: 99%