Learn More
Medchemexpress LLC YKL-5-124 (TFA) | 2748220-93-9 | 99.7% | 629.63 g/mol | C30H34F3N7O5 | 10 MG
SDP
Catalog No. 5000211151
Encompass_Preferred
YKL-5-124 TFA is the trifluoroacetate salt of a potent, selective, irreversible covalent inhibitor of cyclin-dependent kinase 7 (CDK7). It is intended for in vitro and preclinical research into cell-cycle regulation and oncology, supplied in small milligram quantities for biochemical and cellular studies.
  • Potent, selective, irreversible covalent CDK7 inhibitor.
  • Demonstrated biochemical and cellular activity in CDK7 assays.
  • Used to induce cell-cycle arrest and inhibit E2F-driven gene expression.
  • High purity suitable for research use (≈99.7%).
  • Supplied in small-scale quantities for research (e.g., 10 mg).
  • Trifluoroacetate salt; CAS 2748220-93-9; molecular weight 629.63 g/mol.
Catalog No. 50-002-11151 Supplier Medchemexpress LLC Supplier No. HY101257B10MG
YKL-5-124 TFA is the trifluoroacetate salt of a potent, selective, irreversible covalent inhibitor of cyclin-dependent kinase 7 (CDK7). It is intended for in vitro and preclinical research into cell-cycle regulation and oncology, supplied in small milligram quantities for biochemical and cellular studies.
  • Potent, selective, irreversible covalent CDK7 inhibitor.
  • Demonstrated biochemical and cellular activity in CDK7 assays.
  • Used to induce cell-cycle arrest and inhibit E2F-driven gene expression.
  • High purity suitable for research use (≈99.7%).
  • Supplied in small-scale quantities for research (e.g., 10 mg).
  • Trifluoroacetate salt; CAS 2748220-93-9; molecular weight 629.63 g/mol.