Learn More
Medchemexpress LLC YKL-5-124 trifluoroacetate | 2748220-93-9 | 99.7% | 629.63 | C30H34F3N7O5 | 10 MM x 1 ML
SDP
Catalog No. 5000212846
Encompass_Preferred
YKL-5-124 TFA is the trifluoroacetate salt of a potent, selective, irreversible, covalent cyclin-dependent kinase 7 (CDK7) inhibitor. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use in biochemical and cellular assays and is reported to induce cell-cycle arrest and inhibit E2F-driven gene expression while minimally affecting RNA polymerase II phosphorylation.
  • Potent, selective CDK7 inhibition
  • Irreversible covalent mechanism of action
  • Supplied as a 10 mM solution in DMSO, 1 mL vial
  • High purity suitable for biochemical and cellular studies
  • Molecular formula and mass provided for identification
Catalog No. 50-002-12846 Supplier Medchemexpress LLC Supplier No. HY101257B10MM 1ML
YKL-5-124 TFA is the trifluoroacetate salt of a potent, selective, irreversible, covalent cyclin-dependent kinase 7 (CDK7) inhibitor. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use in biochemical and cellular assays and is reported to induce cell-cycle arrest and inhibit E2F-driven gene expression while minimally affecting RNA polymerase II phosphorylation.
  • Potent, selective CDK7 inhibition
  • Irreversible covalent mechanism of action
  • Supplied as a 10 mM solution in DMSO, 1 mL vial
  • High purity suitable for biochemical and cellular studies
  • Molecular formula and mass provided for identification