Learn More
Sino Biological YKL-40 / CHI3L1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11227T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human YKL-40 / CHI3L1 (rh YKL-40 / CHI3L1; Catalog#11227-H08H; NP_001267.2; Met1-Thr383). YKL-40 / CHI3L1 specific IgG was purified by Human YKL-40 / CHI3L1 affinity chromatography.
Catalog No. 50-224-6362