Learn More
Medchemexpress LLC YL-109 | 36341-25-0 | 99.26% | 257.31 | 25 MG
SDP
Catalog No. 5000384068
Encompass_Preferred
YL-109 is an antitumor agent that induces carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. It inhibits breast cancer cell growth and invasiveness.
  • Inhibits cell proliferation, motility, and invasiveness in breast cancer cells.
  • Increases both CHIP mRNA and protein levels in MDA-MB-231 cells.
  • Inhibits both tumor growth and cancer metastasis of breast cancer cells in vivo.
  • Store powder at -20°C for 3 years, 4°C for 2 years.
  • Store in solvent at -80°C for 2 years, -20°C for 1 year.
Catalog No. 50-003-84068 Supplier Medchemexpress LLC Supplier No. 25MGHY18619
Add to Cart
Add to Cart
YL-109 is an antitumor agent that induces carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. It inhibits breast cancer cell growth and invasiveness.
  • Inhibits cell proliferation, motility, and invasiveness in breast cancer cells.
  • Increases both CHIP mRNA and protein levels in MDA-MB-231 cells.
  • Inhibits both tumor growth and cancer metastasis of breast cancer cells in vivo.
  • Store powder at -20°C for 3 years, 4°C for 2 years.
  • Store in solvent at -80°C for 2 years, -20°C for 1 year.