Learn More
Medchemexpress LLC YL-109 | 36341-25-0 | 99.3% | 257.31 | 1 ML
SDP
Catalog No. 5000387843
Encompass_Preferred
YL-109 is an antitumor agent that works by inducing carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. It has demonstrated the ability to inhibit breast cancer cell growth and invasiveness.
  • Inhibits cell proliferation, motility, and invasiveness in breast cancer cells
  • Increases CHIP mRNA and protein levels
  • Suppresses tumor growth and cancer metastasis in vivo
  • Strongly inhibited cell proliferation in MCF-7 and MDA-MB-231 cells
Catalog No. 50-003-87843 Supplier Medchemexpress LLC Supplier No. HY1861910MM/1ML
YL-109 is an antitumor agent that works by inducing carboxyl terminus of Hsp70-interacting protein (CHIP) expression through aryl hydrocarbon receptor (AhR) signaling. It has demonstrated the ability to inhibit breast cancer cell growth and invasiveness.
  • Inhibits cell proliferation, motility, and invasiveness in breast cancer cells
  • Increases CHIP mRNA and protein levels
  • Suppresses tumor growth and cancer metastasis in vivo
  • Strongly inhibited cell proliferation in MCF-7 and MDA-MB-231 cells